Learn More
Cole-Parmer™ Vial Extractor/End Plug Remover for Mid-Size and Large Grinding Vials
Shop All SPEX Certiprep Products
Click to view available options
:
Vial Extractor/End Plug Remover; For use with: Grinding Vial Set (Large, Mid Size), Cr-Free Grinding Vial (Large, Mid Size) of Spex SamplePrep Large-Capacity Freezer/Mill
Description
- Makes it easier to open cold grinding vials immediately after they have been removed from 6870 Large Freezer/Mill.
- Lever helps to remove end plugs from center cylinders of large and mid-size grinding vials.
- Compatible with following large grinding vials: 6801 Large Grinding Vial Set and 6871 Large low-chrome Grinding Vial Set
- Compatible with following mid-size vials when used in conjunction with the 6884 Mid-Size Grinding Vial Adapter: 6881 Mid-Size Grinding Vial Set, 6883 Mid-Size low-chrome Grinding Vial Set and 6885 Mid-Size Poly-Vial Set
- Included in 6870L Large Grinding Vial Accessory Package.
Specifications
Specifications
| Product Type | Replacement Extraction Tool |
| For Use With (Application) | Remove End Plugs From The Center Cylinders of Large and Mid Size Grinding Vials |
| For Use With (Equipment) | Grinding Vial Set (Large, Mid Size), Cr-Free Grinding Vial (Large, Mid Size) of Spex SamplePrep Large-Capacity Freezer/Mill |