Learn More
SPEX™ SamplePrep Geno/Grinder™ Accessory, Grinding Mill
Shop All SPEX Certiprep Products
Click to view available options
For Use With (Equipment):
Titer plates and vial sets of Geno/Grinder
Unit Size:
200g
Description
- Acid washed silica grinding beads
- Bead size ranges from 800-1000μm
- Suitable for basic cell lysis applications using the 2010 Geno/Grinder
- Treated to remove fine particles and contaminates
Specifications
Specifications
| Product Type | Silica Grinding Beads |
| For Use With (Application) | Ideal for basic cell lysis |
| For Use With (Equipment) | Titer plates and vial sets of Geno/Grinder |
| Material | Silica |
| Particle Size | 400 to 600 μm |