Learn More
3M™ Speedglas™ Heat Reflecting Front Panel
Description
- Heat reflective
- Clear
- For use with welding helmet
Specifications
Specifications
| Color | Clear |
| For Use With (Equipment) | Speedglas™ Welding Helmet 9000 |
| Product Type | Heat Reflecting Front |