Learn More
Spectroline™ UV Absorbing Face Shields
Description
- Improve contrast between the fluorescent area and the background by eliminating the “blue haze” interference
- Face shield (UVF–80) provide maximum protection from extended or high intensity UV exposure and adjust to fit the face
Compliance
Face shield meets ANSI specifications and OSHA standards.
Specifications
Specifications
| Product Type | UV-absorbing Faceshield |