Learn More
Spectroline™ UV Absorbing Face Shields
Product Code. 11530917
Encompass
Change view
Click to view available options
:
Face Shield
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11530917 Face Shield
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11530917 Supplier Spectroline™ Supplier No. UVF80
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed for long wearing comfort and protection against most ultraviolet light sources
  • Improve contrast between the fluorescent area and the background by eliminating the “blue haze” interference
  • Face shield (UVF–80) provide maximum protection from extended or high intensity UV exposure and adjust to fit the face
Compliance
Face shield meets ANSI specifications and OSHA standards.
Specifications
Product Type UV-absorbing Faceshield