Learn More
3M™ Aura™ Specialty Respirator, Series 9300
Click to view available options
Filter Efficiency:
FFP1
FFP2
FFP3
With Exhalation Valve:
No
Yes
Color:
Blue
Gray
White
Unit Size:
Pack of 10
Pack of 180
Pack of 20
Description
Fits a wide range of face shapes and sizes
- Low breathing resistance filter technology for easier breathing
- Ingenious three-panel design accommodates greater facial movement during speech for more comfortable wear
- An embossed top panel to reduce fogging of eyewear
- A sculpted nose panel for improved compatibility with eyewear
- An innovative chin tab for ease of donning and adjustment of the respirator on the face
- Supplied in hygienic packaging to help protect the respirator from contamination before use, also allows practical storage and dispensing in the workplace.
- Cool Flow™ valve reduces heat build-up to offer workers comfortable protection - even in hot and humid conditions
Specifications
Specifications
| Assigned Protection Factor (APF) | 4 |
| Certifications/Compliance (Europe) | EN149:2001 + A1:2009 |
| Color Code | Yellow |
| Color | Blue |
| Design Type | Cup/Molded |
| Disposable | Single-use |
| Product Type | Particulate Respirator with Nuisance Organic Vapor Relief |
| Size | Standard |
| With Exhalation Valve | No |
| Filter Efficiency | FFP1 |
| Show More |