Description
- Very strong durability
- Suitable for heavy and long-lasting use
- Tightly woven fibers, high abrasion resistance and low-lint surface
- Outstanding absorption performance
- Large absorption capacity (up to 16 time of own weight)
- Light for smaller leakages and prevention
- Heavy for larger leakages or absorption of puddles
Specifications
Specifications
| Color | Yellow |
| Material | Heavy-duty Cloth |
| Type | Sorbent Sock |
| Length (English) | 9.84 ft. |
| Length (Metric) | 3 m |
| Width (English) | 2.95 in. |
| Width (Metric) | 75.00 mm |
| Weight (English) | 11.90 lb. |
| Weight (Metric) | 5.4 kg |