Learn More
Hellma™ Special Optical Glass Micro Cells
Product Code. 13494676
Click to view available options
Capacity (Metric):
0.5 mL
Unit Size:
Each
Product Code. 13494676
Brand: Hellma™ 1080021040
Only null left
Specifications
Type Micro Cell
Capacity (Metric) 0.5 mL
Material Special Optical Glass
Width (Metric) Interior 2mm
Quantity 2Pack
Path Length 10 mm
Wavelength Range 200 to 2500 nm
Height (Metric) 45 mm
Width (Metric) 12.5 mm