Learn More
Spautin-1, MedChemExpress
Product Code. 16488177
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488177 5 mg 5mg
16445468 10 mg 10mg
16421647 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16488177 Supplier Medchem Express Supplier No. HY12990
MedChemExpress Spautin-1 is a specific and potent autophagy inhibitor which inhibits ubiquitin-specific peptidases, USP10 and USP13 with IC50s of 0.6-0.7 μM.
Spautin-1 is a specific and potent autophagy inhibitor which inhibits ubiquitin-specific peptidases, USP10 and USP13 with IC50s of 0.6-0.7 μM.
Specifications
Chemical Name or Material Spautin-1
CAS 1262888-28-7
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C15H11F2N3
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (184.32 mM; Need ultrasonic)
SMILES FC1=CC2=C(N=CN=C2C=C1)NCC3=CC=C(F)C=C3
Molecular Weight (g/mol) 271.26
Formula Weight 271.26
Percent Purity 99.26%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death