Learn More
BRAND™ Spare Lateral Stopcock with Key
Stopcock housing with burette tip includes interchangeable PTFE or borosilicate glass key with retention device.
20.35 EUR - 40.45 EUR
Specifications
Capacity (Metric) | 50 mL |
---|---|
For Use With (Equipment) | Burette |
No. per Pack | 1 Each |
Includes | Retention Device |
Type | Stopcock Key |
Product Code | Brand | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Price | Quantity & Availability | |||||
10278401
|
BRAND™
81167 |
PTFE |
40.45 EUR
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
10750043
|
BRAND™
81142 |
Borosilicate Glass |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Lateral stopcock housing with burette tip
- Interchangeable PTFE or borosilicate glass key with retention device
Specifications
50 mL | |
1 Each | |
Stopcock Key |
Burette | |
Retention Device |