Learn More
Sorbic acid potassium salt, ∽99%, MP Biomedicals™
Product Code. 11426161 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 g
500 g
Unit Size:
250g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11426161 250 g 250g
11436161 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11426161 Supplier MP Biomedicals Supplier No. 0219114983
Chemical Identifiers
CAS 590-00-1
Molecular Formula C6H7KO2
Molecular Weight (g/mol) 150.22
MDL Number MFCD00016546
InChI Key CHHHXKFHOYLYRE-STWYSWDKSA-M
Synonym potassium sorbate, sorbistat-k, potassium 2,4-hexadienoate, sorbistat potassium, potassium e,e-sorbate, sorbic acid potassium salt, potassium 2e,4e-hexa-2,4-dienoate, bb powder, sorbistat-potassium, caswell no. 701c
PubChem CID 23676745
ChEBI CHEBI:77868
IUPAC Name potassium (2E,4E)-hexa-2,4-dienoate
SMILES [K+].C\C=C\C=C\C([O-])=O
Specifications
Color White
Density 1.36g/mL
Assay Percent Range 99%
Quantity 250 g
Formula Weight 150.2
Physical Form Crystalline Powder
Chemical Name or Material Sorbic acid potassium salt