missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Soot Content Standard Set (contains one of each of the following: SOOT-BLK, SOOT-A, SOOT-B, SOOT-C, SOOT-D, SOOT-E), VHG™
Description
Soot Content Standard Set (contains one of each of the following: SOOT-BLK, SOOT-A, SOOT-B, SOOT-C, SOOT-D, SOOT-E)
Specifications
Specifications
| Methods | FTIR/IR |
| Shipping Condition | Room Temperature |
| Solvent or Matrix | 15W40 Motor Oil |
| Quantity | 6 x 50 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?