missing translation for 'onlineSavingsMsg'
Learn More

Solutol HS-15 (Technical Grade), TRC

Product Code. 30424960
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424960 100 mg 100mg
30427178 500 mg -
30413059 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30424960 Supplier Toronto Research Chemicals Supplier No. TRCS676545100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61909-81-7
Molecular Formula (C2H4O)nC18H36O3
Molecular Weight (g/mol) 344.5
Synonym Polyethylene Glycol 12-hydroxystearate, Kolliphor HS 15, Solutol HS 15
IUPAC Name 13-hydroxy-1-(2-hydroxyethoxy)nonadecan-2-one
SMILES CCCCCCC(O)CCCCCCCCCCC(=O)COCCO

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C21H42O4/c1-2-3-4-11-14-20(23)15-12-9-7-5-6-8-10-13-16-21(24)19-25-18-17-22/h20,22-23H,2-19H2,1H3
Formula Weight 358.3083
Chemical Name or Material Solutol HS-15 (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.