Learn More
3M™ Solus™ Anti-Fog Coating AF-AS lens
Click to view available options
Temple Color:
Gray/Blue-Green
Gray/Red
Lens Tint:
Clear
Gray
Unit Size:
Pack of 20
Description
- Adjustable ratchet temples
- Scotchgard™ Anti-Fog Coating resists fogging longer than traditional anti-fog coatings, even after up to 25 washings with water
- 3M internal testing per EN168 test method when compared with traditional anti-fog coatings.
- Polycarbonate lens absorb 99.9% of UVA and UVB rays
- Meets the requirements of EN166-2001 Standard as an Impact Rated Spectacle
Specifications
Specifications
| Lens Coating | Anti-fog, Scratch-resistant |
| Lens Tint | Clear |
| Frame Color | Gray |
| Quantity | 20/Cs. |
| Temple Color | Gray/Blue-Green |
| Lens Material | Polycarbonate |
| Frame Material | Plastic |
| Frame Type | Half Frame |