Learn More
Fisherbrand™ Solid Natural Rubber Stopper
Shop All Fisherbrand Products
missing translation for 'orderingAttributeHoverText'
Diameter (Metric) Bottom:
79 mm
92 mm
missing translation for 'unitSize'
Each
Description
- Small, tapered plug used to seal the openings of test tubes, flasks and other laboratory glassware
- Preferable for applications that require tighter seal or greater degree of chemical resistance
- Prevent contamination of samples by protecting contents of laboratory glassware from air
- Allows experimenters to shake or mix solutions without spilling them
Specifications
Specifications
| Hardness | 46/55 IRHD |
| Height (Metric) | 54mm |
| Quantity | 1/Ea. |
| Color | Red |
| Diameter (Metric) Bottom | 79 mm |
| Material | Natural Rubber |
| Product Type | Solid Stopper |