missing translation for 'onlineSavingsMsg'
Learn More

Sograzepide, MedChemExpress

Product Code. 16476566
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476566 100 mg 100mg
16478797 5 mg 5mg
16452337 50 mg 50mg
16436208 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16476566 Supplier Medchem Express Supplier No. HY14850

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sograzepide (Netazepide; YF 476; YM-220) is an extremely potent , highly selective and orally active Gastrin/CCK-B antagonist with an IC50 value of 0.1 nM, has inhibitory effect on Gastrin/CCK-A activity with an IC50 of 502 nM.

Sograzepide (Netazepide; YF 476; YM-220) replaces the specific binding of [125I]CCK-8 to the rat brain, cloned canine and cloned human Gastrin/CCK-B receptors, with Ki values of 0.068, 0.62 and 0.19 nM, respectively.

Specifications

Chemical Name or Material Sograzepide
Physical Form Solid
CAS 155488-25-8
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C28H30N6O3
For Use With (Application) Metabolism-protein/nucleotide metabolism
Synonym Netazepide YF 476 YM-220
Solubility Information DMSO : ≥ 100 mg/mL (200.57 mM)
SMILES O=C(NC1=CC=CC(NC)=C1)N[C@H]2C(N(CC(C(C)(C)C)=O)C3=CC=CC=C3C(C4=NC=CC=C4)=N2)=O
Molecular Weight (g/mol) 498.58
Formula Weight 498.58
Percent Purity 98.35%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.