missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate, 13% in water, EssentQ, Scharlab

Product Code. 11338584
Change view
Click to view available options
Quantity:
125 mL
Unit Size:
1 ampoule
Product Code. Quantity unitSize
11338584 125 mL 1 ampoule
1 options
This item is not returnable. View return policy

Product Code. 11338584

Brand: Scharlab SO072800PA

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 10102-17-7
Quantity 125 mL
Molecular Formula H10Na2O8S2
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
SMILES O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
IUPAC Name disodium pentahydrate sulfanidesulfonate
Molecular Weight (g/mol) 248.17
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.