missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium tauroglycocholate, MedChemExpress
Description
Sodium tauroglycocholate is an inhibitor of the biliary acid transporting system of the hepatocyte and also a surfactant used as a chemical permeation enhancer.
Specifications
Specifications
| Chemical Name or Material | Sodium tauroglycocholate |
| CAS | 41945-48-6 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C28H47N2NaO8S |
| Quantity | 100 mg |
| Synonym | Tauroglycocholic acid sodium salt |
| Solubility Information | DMSO : 30 mg/mL (50.44 mM; Need ultrasonic and warming) |
| SMILES | O[C@H](C[C@@](C[C@H](O)CC1)([H])[C@@]1(C)[C@@]2([H])C[C@@H]3O)[C@@]2([H])[C@@](CC[C@]4([H])[C@H](C)CCC(NCC(NCCS(=O)(O[Na])=O)=O)=O)([H])[C@]34C |
| Molecular Weight (g/mol) | 594.74 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?