missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfate, TRC

Product Code. 17998504
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17998504 10 g 10g
17969444 1 g 1g
17938514 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17998504 Supplier Toronto Research Chemicals Supplier No. TRCS67362010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-82-6
Molecular Formula 2 Na . O4 S
Molecular Weight (g/mol) 142.04
Synonym Sulfuric acid sodium salt (1:2), Sulfuric acid disodium salt (8CI,9CI), Sodium sulfate (2:1), A 12, A 12 (sulfate), A 6, A 6 (salt), Bisodium sulfate, Dibasic sodium sulfate, Disodium monosulfate, Disodium sulfate, Disodium sulphate, Kemsol, Na-G 0, Na-G 1, Salt cake, Sodium sulfate, Sodium sulfate (Na2SO4), Sodium sulfate anhydrous, Sodium sulphate, Sulfuric acid, disodium salt, T 350, T 350 (sulfate)
IUPAC Name disodium;sulfate
SMILES [Na+].[Na+].[O-]S(=O)(=O)[O-]

Specifications

Quantity 10 g
InChI Formula InChI=1S/2Na.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2
Formula Weight 141.9313
Chemical Name or Material Sodium Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.