missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium nitrite, reagent grade, Scharlab
Click to view available options
Quantity:
500 g
Unit Size:
500g
Specifications
Specifications
| CAS | 7632-00-0 |
| Quantity | 500 g |
| Molecular Formula | NNaO2 |
| InChI Key | LPXPTNMVRIOKMN-UHFFFAOYSA-M |
| SMILES | [Na+].[O-]N=O |
| IUPAC Name | sodium nitrite |
| Molecular Weight (g/mol) | 69.00 |
| Grade | ExpertQ |
Safety and Handling
Product Identifier
- Sodium nitrite, reagent grade, ACS
- Danger
- Oxidising solid Category 3
- Acute toxicity Category 3
- ACUTE AQUATIC Acute 1
- H272-May intensify fire; oxidiser.
- H301-Toxic if swallowed.
- H400-Very toxic to aquatic life.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501-Dispose of contents/container to .
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur