Learn More
Sodium nicotinate, 98%
Product Code. 11415866
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Product Code. Quantity unitSize
11415866 100 g 100g
11425866 500 g 500g
2 options
Product Code. 11415866
Brand: Thermo Scientific Alfa Aesar B22224.22
Request Bulk or Custom Format
CAS: 54-86-4 | C6H4NNaO2 | 145.093 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 54-86-4
Molecular Formula C6H4NNaO2
Molecular Weight (g/mol) 145.093
MDL Number MFCD00064352
InChI Key KFLRWGSAMLBHBV-UHFFFAOYSA-M
Synonym sodium nicotinate, nicotinic acid sodium salt, sodium pyridine-3-carboxylate, natriumnicotinat german, unii-3zx78b4q2l, natriumnicotinat, 3-pyridinecarboxylic acid, sodium salt, nicotinic acid, sodium salt, 3-pyridinecarboxylic acid, sodium salt 1:1, nicotinic acid sodium
PubChem CID 23687810
IUPAC Name sodium;pyridine-3-carboxylate
SMILES C1=CC(=CN=C1)C(=O)[O-].[Na+]
Specifications
Melting Point >300°C
Quantity 100 g
Formula Weight 145.1
Percent Purity 98%
Chemical Name or Material Sodium nicotinate