missing translation for 'onlineSavingsMsg'
Learn More

Sodium ionophore III, MedChemExpress

Product Code. 16404536
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
This item is not returnable. View return policy

Product Code. 16404536

Brand: Medchem Express HY101109

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sodium ionophore III (ETH2120) is a Na+ ionophore suitable for the assay of sodium activity in blood, plasma, serum. etc.

Sodium ionophore III (ETH2120) is a Na+ ionophore suitable for the assay of sodium activity in blood, plasma, serum. etc.

Specifications

Chemical Name or Material Sodium ionophore III
CAS 81686-22-8
Color White
Purity Grade Notes Research
Molecular Formula C34H52N2O4
Quantity 100 mg
Synonym ETH2120
Solubility Information DMSO : 2 mg/mL (3.62 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 1 mg/mL (1.81 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C(N(C1CCCCC1)C2CCCCC2)COC3=CC=CC=C3OCC(N(C4CCCCC4)C5CCCCC5)=O
Molecular Weight (g/mol) 552.79
Formula Weight 552.79
Percent Purity 98.32%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.