missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 5M, Chem Lab™

Product Code. 11923253
Click to view available options
Quantity:
5 L
10 L
Packaging:
473mL
10L
Unit Size:
10 L
5 L
This item is not returnable. View return policy

Product Code. 11923253

Brand: Chem-Lab CL05.1435.9010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Color Colorless
Concentration 14.16g NaOH/l H2O = 1/2.825 N (±0.0007/20°C)
Density 1.02g/mL
Solubility Information Soluble in water
Quantity 10 L
Odor Odourless
Packaging 10L
Physical Form Liquid
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Sodium hydroxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.