missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 50% w/v, Extra Pure, Scharlab

Product Code. 11338524
Change view
Click to view available options
Quantity:
1 L
Packaging:
HDPE Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11338524 1 L HDPE Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11338524 Supplier Scharlab Supplier No. SO04241000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Suitable for qualitative and semi-quantitative work. Sodium Hydroxide, 50% w/v, Extra Pure, Scharlab is often used in organic chemistry for purifying synthesized products when higher grade solvents than Synthesis Grade are required.

Extra Pure grade specifications are less strict than Reagent Grade, but are significantly higher than industrial products. They are often used in the pharmaceutical industry, either as a reagent or as raw materials for production.

Specifications

Color Colorless
Boiling Point 143°C
Melting Point 12°C
CAS 1310-73-2
Molecular Formula NaOH
UN Number 1824
Density 1.4g/cm3
Synonym Sodium oxidanide
Solubility Information Miscible in water
Quantity 1 L
Formula Weight 40.00g/mol
Percent Purity ≥50%
Grade Applications: analytical chemistry, laboratory reagent, solvents, for pharmaceutical use, for the synthesis of inorganic salts
Packaging HDPE Bottle
Vapor Pressure 2 hPa
Physical Form Solution
Assay w/v, acidimetric
Chemical Name or Material Sodium hydroxide
Show More Show Less
compliance-icons
Product Identifier
  • Sodium hydroxide, solution 50% w/v, extra pure
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: Sodium Hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.