missing translation for 'onlineSavingsMsg'
Learn More

Sodium Fluoroacetate, MP Biomedicals

Product Code. 11471795 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11471795 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11471795 Supplier MP Biomedicals Supplier No. 0520108001

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 62-74-8
Molecular Formula C2H2FNaO2
MDL Number MFCD00002682
Quantity 1 g
Synonym sodium fluoroacetate, sodium monofluoroacetate, furatol, yasoknock, compound 1080, fratol, tenate, fluoroacetic acid sodium salt, natriumfluoracetat, fluorakil 3
InChI Key JGFYQVQAXANWJU-UHFFFAOYSA-M
SMILES [Na+].[O-]C(=O)CF
IUPAC Name sodium;2-fluoroacetate
Molecular Weight (g/mol) 100.02
PubChem CID 16212360
ChEBI CHEBI:38699
Formula Weight 100.02
Chemical Name or Material Sodium Fluoroacetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.