missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Packaging | Bottle |
| CAS | 497-19-8, 7732-18-5 |
| Molecular Formula | CNa2O3 |
| Concentration | 0.05 M |
| MDL Number | MFCD00003494 |
| Synonym | sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda |
| InChI Key | CDBYLPFSWZWCQE-UHFFFAOYSA-L |
| SMILES | [Na+].[Na+].[O-]C([O-])=O |
| IUPAC Name | disodium carbonate |
| Molecular Weight (g/mol) | 105.99 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?