Learn More
Sodium Acetate Trihydrate, MP Biomedicals
Product Code. 11495735 Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 g
1 kg
6 x 1 kg
Unit Size:
1kg
500g
6kg
Product Code. 11495735
Brand: MP Biomedicals 0230018406
  • To 800-900 mL deionized water add: 8.20 g sodium acetate and mix
  • Adjust to pH 5.2 with 1.0 M NaOH
  • QS to 1 L with deionized water
  • Mix
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6131-90-4
Molecular Formula C2H9NaO5
Molecular Weight (g/mol) 136.079
MDL Number MFCD00071557
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
Synonym sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
PubChem CID 23665404
ChEBI CHEBI:32138
IUPAC Name sodium;acetate;trihydrate
SMILES CC(=O)[O-].O.O.O.[Na+]
Specifications
Melting Point 56°C to 64°C (literature)
Color White
Boiling Point 123°C (-3H)
Linear Formula CH3COONa . 3H2O
Quantity 6 x 1 kg
Solubility Information Soluble in water (20 g plus 150 mL).(Lit.)
Formula Weight 136.08
Grade HPLC
Physical Form Crystalline Powder
Chemical Name or Material SODIUM ACETATE TRIHYDRATE