missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate, 1M aq. soln., pH 4.5, RNAse free

Product Code. p-7042760
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
This item is not returnable. View return policy

Product Code. 15450317

Brand: Thermo Scientific Alfa Aesar J63669.AK

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol

Sodium Acetate is used as a buffer in the buffering range of pH 3.6-5.6. And also used in the purification and precipitation of nucleic acids, in protein crystallization, Staining of gels in protein gel electrophoresis and HPLC. Mordant dyeing; DNA microarray studies of E. coli response. Investigation of protein unfolding during reverse phase chromatography. Used in conjunction with sodium carbonate to enhance the activation of freeze-dried subtilisin Carlsberg in organic solvents. As a substrate of acetokinase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium Acetate is used as a buffer in the buffering range of pH 3.6-5.6. And also used in the purification and precipitation of nucleic acids, in protein crystallization, Staining of gels in protein gel electrophoresis and HPLC. Mordant dyeing; DNA microarray studies of E. coli response. Investigation of protein unfolding during reverse phase chromatography. Used in conjunction with sodium carbonate to enhance the activation of freeze-dried subtilisin Carlsberg in organic solvents. As a substrate of acetokinase.

Solubility
Fully miscible.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
PubChem CID 517045
ChEBI CHEBI:32954
SMILES [Na+].CC([O-])=O

Specifications

pH 4.5
Physical Form Liquid
Quantity 250 mL
Linear Formula NaOOCCH3
Solubility Information Fully miscible.
IUPAC Name sodium acetate
Formula Weight 82.03
Concentration or Composition (by Analyte or Components) 1M aq. soln.
Odor Odorless
Chemical Name or Material Sodium acetate, RNAse free

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.