Learn More
Sodium 140/Potassium 5mmol/L, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Specifications
Specifications
| Certificate of Analysis | NIST Certificate |
| Concentration | 5/40m mol/L |
| Quantity | 500 mL |