missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-Hexanesulfonate Monohydrate, 99%

Product Code. p-7049966
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368337 10 g 10g
11358337 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368337 Supplier Thermo Scientific Alfa Aesar Supplier No. 032190.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 207300-91-2 | C6H15NaO4S | 206.232 g/mol

Sodium 1-hexanesulfonate monohydrate is an ion pair reagent, which is used for the analysis of small organic compounds, pharmaceutical products and metabolites using high-performance liquid chromatography (HPLC). It is an efficient catalyst for the preparation of alpha-aminophosphonates by the coupling of aldehydes/ketone, an amine and triethyl phosphate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207300-91-2
Molecular Formula C6H15NaO4S
Molecular Weight (g/mol) 206.232
MDL Number MFCD00149549
InChI Key CLCGFJYKZGFGSQ-UHFFFAOYSA-M
Synonym 1-hexanesulfonic acid sodium salt monohydrate, sodium hexane-1-sulfonate hydrate, sodium 1-hexanesulfonate monohydrate, n-1-hexanesulfonic acid, sodium 1-hexanesulfonate hydrate, sodium n-hexanesulfonate monohydrate, potassium hexane-1-sulfonate hydrate, sodium hexane-1-sulphonate monohydrate, sodium hexane-1-sulfonate-water 1/1/1, 1-hexanesulfonic acid,sodium salt, hydrate 1:1:1
PubChem CID 23696962
IUPAC Name sodium;hexane-1-sulfonate;hydrate
SMILES CCCCCCS(=O)(=O)[O-].O.[Na+]

Specifications

Color White
Linear Formula CH3(CH2)5SO3Na·H2O
Quantity 10 g
Beilstein 3727014
Solubility Information Slightly soluble in water
Formula Weight 206.24 (188.22 Anhydrous)
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Sodium 1-hexanesulfonate monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.