Learn More
DWK Life Sciences Kimble Chase™ Soda Lime Glass Tube
Product Code. 11862383
Click to view available options
Diameter (Metric):
19 mm
21 mm
22 mm
24 mm
25 mm
27 mm
28 mm
30 mm
Length (Metric):
40 mm
45 mm
50 mm
52 mm
55 mm
70 mm
75 mm
80 mm
100 mm
Unit Size:
Pack of 200
Product Code. 11862383
Brand: DWK Life Sciences F40452200B0W2
Only null left
Add to basket
Add to basket
Conduct research and experimentation with Soda Lime Glass Tube. This basic test tube has an open end, round bottom and a cap.
  • Reusable
  • 200Pack
  • Clear soda lime glass
  • With cap
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 45 mm
Packaging 200Pack
Diameter (Metric) 22 mm
Product Type Test Tube