Learn More
Fisherbrand™ Soda Lime Glass Light Wall Rimmed Test Tube
Product Code. 15832265 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric):
10 mm
12 mm
16 mm
24 mm
Length (Metric):
75 mm
100 mm
125 mm
150 mm
Unit Size:
Pack of 100
Pack of 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Length (Metric) unitSize
15832265 12 mm 100 mm Pack of 100
15892255 10 mm 75 mm Pack of 100
15852265 16 mm 125 mm Pack of 100
15872265 16 mm 150 mm Pack of 100
15812265 12 mm 75 mm Pack of 100
15842275 24 mm 150 mm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Product Code. 15832265
Only null left
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Manufactured from soda lime glass and constructed with light walls. Fisherbrand™ Soda Lime Glass Light Wall Rimmed Test Tubes are ideal for general mixing and laboratory work.
  • Withstands moderate warming
  • Light weight
  • Complies with ISO 4142
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 4142
Length (Metric) 100 mm
Material Soda Lime Glass
Quantity 100 Pack
Rimmed Yes
Diameter (Metric) 12 mm