Learn More
Fisherbrand™ Soda Lime Glass Funnel
Product Code. p-8000436 Shop All Fisherbrand Products
Click to view available options
Diameter (Metric)Top:
45mm
50mm
55mm
60mm
75mm
100mm
Length (Metric) Stem:
45mm
50mm
55mm
60mm
70mm
100mm
Unit Size:
Each
Product Code. 11562423
Only null left
Add to basket
Add to basket
Plain glass funnel manufactured from soda lime glass. Fisherbrand™ Soda Lime Glass Funnel is ideal for use with funneling liquids in laboratory applications.
Plain glass funnel manufactured from soda lime glass.
Specifications
Diameter (Metric)Top 100mm
Material Soda Lime Glass
Length (Metric) Stem 100mm