missing translation for 'onlineSavingsMsg'
Learn More

SNX9 Monoclonal Antibody (OTI1E2), TrueMAB™, OriGene

Product Code. 17593701
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17593701 30 μL 30µL
17573701 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17593701 Supplier Origene Technologies Supplier No. TA501250S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SNX9 Monoclonal antibody specifically detects SNX9 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen SNX9
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1E2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ARNT
Gene Accession No. P27540
Gene Alias 2700073N08Rik; MST155; MSTP155; OTTHUMP00000040083; Protein SDP1; RP11-266C7.10; SDP1; SH3 and PX domain-containing protein 1; SH3 and PX domain-containing protein 3A; SH3PX1; SH3PXD3A; Snx9; sorting nexin 9; sorting nexin-9; Wiskott-Aldrich syndrome protein (WASP) interactor protein; WISP
Host Species Mouse
Immunogen Full length human recombinant protein of human ARNT produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 405
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.