missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNAP25 Monoclonal Antibody (SP12)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA517609
This item is not returnable.
View return policy
Description
MA5-17609 targets SNAP-25 in ELISA, FACS, and WB applications and shows reactivity with Human samples. The MA5-17609 immunogen is human Synaptobrevin 1 (aa 1-118) and Synaptobrevin 2(aa 33-96).
SNAP25 is a t-SNARE involved in the molecular regulation of neurotransmitter release. It may play a role in the synaptic function of specific neuronal systems. SNAP25 associates with proteins involved in vesicle docking and membrane fusion. It also regulates plasma membrane recycling through its interaction with CENPF. Mutations in the gene can results in myasthenic syndrome.
Specifications
| SNAP25 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P60879, P60880, P60881 | |
| Snap25 | |
| Synaptic preparation of post mortem human brain. | |
| 200 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
| SP12 | |
| Unconjugated | |
| Snap25 | |
| ALMS1; bA416N4.2; Bdr; blind drunk; CG17676; CG17884; CG40452; CMS18; dJ1068F16.2; Dmel CG40452; Dmel_CG40452; etID17571.20; etID69167.20; fj42b02; fj45g04; fj48c02; FLJ23079; GENA70; HGNC:11132; resistance to inhibitors of cholinesterase 4 homolog; RIC4; RIC-4; SEC9; si:dkeyp-8f4.6; Snap; SNAP 25; snap25; SNAP-25; snap25.1; SNAP-25.1; snap25a; SNAP-25A; SNAP-25B; snp25; sp; SUP; super protein; synaptosomal associated protein-25; synaptosomal-associated 25 kDa protein; Synaptosomal-associated protein 25; synaptosomal-associated protein 25-A; synaptosomal-associated protein, 25 kDa; synaptosomal-associated protein, 25a; synaptosomal-associated protein, 25kDa; synaptosome associated protein 25; synaptosome associated protein 25a; synaptosome associated protein 25kDa; synaptosome-associated protein 25.1; synaptosome-associated protein 25a; wu:fj42b02; wu:fj45g04; wu:fj48c02 | |
| Mouse | |
| Protein G | |
| RUO | |
| 20614, 25012, 6616 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction