Få mere at vide
Invitrogen™ Snail1 Monoclonal Antibody (20C8), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
165.00 EUR - 3585.00 EUR
Tekniske data
| Antigen | Snail1 |
|---|---|
| Clone | 20C8 |
| Concentration | 0.5 mg/mL |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Produktkode | Brand | Quantity | Pris | Mængde & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Quantity | Pris | Mængde & tilgængelighed | |||||
|
15537696
|
Invitrogen™
14-9859-80 |
25 μg |
165.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17110812
|
Invitrogen™
14-9859-37 |
2 mg |
1586.00 EUR
2mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17120812
|
Affymetrix eBioscience
14-9859-95 |
10 mg | |||||||
Beskrivelse
The MAb 20C8 recognizes human Snail1, a 29 kDa zinc finger transcription factor. Snail1 and Slug both belong to a family of transcription factors that are responsible for inducing epithelial to mesenchymal transition (EMT) during normal organ development and tumor progression. Snail1 expression is localized to both the cytoplasm and nucleus and functions as a transcriptional repressor of E-cadherin, claudins, occludin, and desmoplakin. During embryonic development, Snail1 is localized to the mesoderm, while in the adult, expression is inducible and is usually limited to activated fibroblasts and some mesenchymal cells. Overexpression of Snail1 has been found in carcinomas and fibrosarcomas and may be used as an early marker for EMT.
Zinc finger protein SNAI1 (SNAIL)is a zinc finger transcriptional repressor which down regulates the expression of the ectodermal gene within the mesoderm and functions to form the mesoderm and neural crest. It is reported that SNAI1 acts as an important factor of tumor invasion as evidenced by its role in E-cadherin down-regulation and induction of epithelial-mesenchymal transition (EMT). In human breast cancer, the expression of SNAI1 and/or the homologous SNAI2 (Slug) has been associated with E-cadherin repression, local or distant metastasis, tumor recurrence, or poor prognosis in different tumor series. It is also reported that Snail 1 protein in the stromamay be anew putative prognosis marker for colon tumors.Tekniske data
| Snail1 | |
| 0.5 mg/mL | |
| Unconjugated | |
| Mouse | |
| Human | |
| O95863 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
| 20C8 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| 6615 | |
| Primary | |
| 4°C | |
| Snai1 |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.