missing translation for 'onlineSavingsMsg'
Learn More

Sn @ 10μg/mL; Si @ 30μg/mL; Mo @ 50μg/mL in 3% HNO3 (v/v)/tr HF, VHG™

Product Code. 17987919
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17987919 500 mL 500mL
17948809 100 mL 100mL
17967359 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 17987919 Supplier VHG Labs Supplier No. VHG1204500

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Sn @ 10μg/mL; Si @ 30μg/mL; Mo @ 50μg/mL in 3% HNO3 (v/v)/tr HF
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tin/Silicon/Molybdenum
CAS 7440-31-5, 7440-21-3, 7439-98-7
Analyte or Component Names Tin/Silicon/Molybdenum
Shipping Condition Room Temperature
Solvent or Matrix 3% HNO3
Quantity 500 mL
Concentration or Composition (by Analyte or Components) Tin 10 μg/mL/Silicon 30 μg/mL/Molybdenum 50 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.