Learn More
SMI-16a, MedChemExpress
Product Code. 16466387
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16466387 5 mg 5mg
16440047 50 mg 50mg
16412816 25 mg 25mg
16403658 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16466387 Supplier Medchem Express Supplier No. HY101947
MedChemExpress SMI-16a is a selective Pim kinase inhibitor with IC50 values of 0.15, 0.02 and 48 μM for Pim1, Pim2 and PC3 cells, respectively.
SMI-16a is a selective Pim kinase inhibitor with IC50 values of 0.15, 0.02 and 48 μM for Pim1, Pim2 and PC3 cells, respectively.
Specifications
Chemical Name or Material SMI-16a
CAS 587852-28-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C13H13NO3S
Quantity 5 mg
Synonym PIM1/2 Kinase Inhibitor VI
Solubility Information DMSO : ≥ 100 mg/mL (379.78 mM)
SMILES O=C(NC/1=O)SC1=C\C2=CC=C(OCCC)C=C2
Molecular Weight (g/mol) 263.31
Formula Weight 263.31
Percent Purity 99.7%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease