missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMG1 Rabbit anti-Human, Mouse, Rat, Clone: 5N10C7, Novus Biologicals™
Rabbit Monoclonal Antibody
190.00 EUR - 496.00 EUR
Specifications
| Antigen | SMG1 |
|---|---|
| Clone | 5N10C7 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18375056
|
Novus Biologicals
NBP3-15716-20UL |
20 μg |
190.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18390864
|
Novus Biologicals
NBP3-15716-100UL |
100 μg |
496.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SMG1 Monoclonal antibody specifically detects SMG1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| SMG1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 23049 | |
| IgG | |
| Affinity purified |
| 5N10C7 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ATXserine/threonine-protein kinase SMG1, EC 2.7.11, EC 2.7.11.1, hSMG-1,61E3.4, KIAA0421phosphatidylinositol 3-kinase-related protein kinase, Lambda/iota protein kinase C-interacting protein, Lambda-interacting protein, LIPPI-3-kinase-related kinase SMG-1, phosphatidylinositol 3-kinase-related kinase, SMG-1, SMG1 homolog, phosphatidylinositol 3-kinase-related kinase (C. elegans) | |
| A synthetic peptide corresponding to a sequence within amino acids 3562-3661 of human SMG1 (Q96Q15). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title