missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SMEK1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16559921
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16559921 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16559921 Supplier Invitrogen™ Supplier No. PA597089

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: VQTFKGLKLR FEQQRERQDN PKLDSMRSIL RNHRYRRDAR TLEDEEEMWF NTDEDDMEDG EAVVSPSDKT KNDDDIMDPI SKFMERKKLK ESEEKEVLLK TNLSGRQSPS FKLSLSSGTK TNLTSQSSTT NLPGSPGSPG SPGSPGSPGS VPKNTSQTAA ITTKGGLVGL VDYPDDDEDD DEDEDKEDTL PLSKKAKFDS; Positive Samples: HeLa, HepG2, Jurkat, MCF-7, Mouse spleen, Rat brain; Cellular Location: Cytoplasm, Nucleus, centrosome, cytoskeleton, microtubule organizing center.

Regulatory subunit of serine/threonine-protein phosphatase 4. May regulate the activity of PPP4C at centrosomal microtubule organizing centers. The PPP4C-PPP4R2-PPP4R3A PP4 complex specifically dephosphorylates H2AFX phosphorylated on 'Ser-140' (gamma-H2AFX) generated during DNA replication and required for DNA DSB repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMEK1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PPP4R3A
Gene Accession No. Q6IN85, Q6P2K6
Gene Alias 1110034C04Rik; BC064465; FLFL1; KIAA2010; mKIAA2010; MSTP033; PP4R3; PP4R3A; Ppp4r3a; protein phosphatase 4 regulatory subunit 3A; protein phosphatase 4, regulatory subunit 3A; RGD1309059; serine/threonine-protein phosphatase 4 regulatory subunit 3A; SMEK homolog 1; SMEK homolog 1, suppressor of mek1; SMEK homolog 1, suppressor of mek1 (Dictyostelium); SMEK1; smk1; smk-1
Gene Symbols PPP4R3A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 395-594 of human SMEK1 (NP_0012712101).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 314388, 55671, 68734
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.