Learn More
Invitrogen™ SMARCA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143864
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: rat testicular issue, mouse testicula issue, human HL-60 whole cell, human THP-1 whole cell, human A549whole cell, human U2OS whole cell, human Hela whole cell. Flow: HeLa cell, THP-1 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
SMARCA2 encodes a protein that is a member of the SWI/SNF family of proteins and is highly similar to the brahma protein of Drosophila. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI, which is required for transcriptional activation of genes normally repressed by chromatin. Two transcript variants encoding different isoforms have been found for this gene, which contains a trinucleotide repeat (CAG) length polymorphism.
Specifications
| SMARCA2 | |
| Polyclonal | |
| Unconjugated | |
| SMARCA2 | |
| 2610209L14Rik; ATP-dependent helicase SMARCA2; BAF190; Baf190b; brahma; BRG1-associated factor 190B; BRM; FLJ36757; global transcription activator homologous sequence; hBRM; hSNF2a; MGC74511; NCBRS; Probable global transcription activator SNF2L2; protein brahma homolog; SMARCA2; SNF2; SNF2/SWI2-like protein 2; SNF2A; SNF2alpha; SNF2-alpha; Snf2l2; SNF2LA; Sth1p; subunit of the SWI/SNF chromatin remodeling complex; sucrose nonfermenting 2-like protein 2; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 2; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin a2; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 2; SWI2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 361745, 6595, 67155 | |
| -20°C | |
| Lyophilized |
| ELISA, Flow Cytometry, Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| P51531, Q6DIC0 | |
| SMARCA2 | |
| E.coli-derived human SMARCA2/BRM recombinant protein (Position: Q181-N624). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.