missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smad7 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99045FR
This item is not returnable.
View return policy
Description
Smad7 Polyclonal antibody specifically detects Smad7 in Human samples. It is validated for Western Blot
Specifications
| Smad7 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 680 | |
| CRCS3, FLJ16482, hSMAD7, MAD homolog 7, MAD homolog 8, MAD, mothers against decapentaplegic homolog 7 (Drosophila), MADH7mothers against decapentaplegic homolog 7, MADH8, Mothers against decapentaplegic homolog 8, Mothers against decapentaplegic, drosophila, homolog of, 7, Mothers against DPP homolog 7, Mothers against DPP homolog 8, SMAD 7, SMAD family member 7MAD (mothers against decapentaplegic, Drosophila) homolog 7, SMAD, mothers against DPP homolog 7, SMAD, mothers against DPP homolog 7 (Drosophila), Smad7 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Smad7. | |
| 0.1 mL | |
| Cell Biology, Signal Transduction | |
| 4092 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction