missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SM68 Standard 3: Au, Ir, Os, Pd, Pt, Rh, Ru, Te @ 100 μg/mL in 10% HCl, VHG™
Description
SM68 Standard 3: Au, Ir, Os, Pd, Pt, Rh, Ru, Te @ 100 μg/mL in 10% HCl
Specifications
Specifications
| Chemical Name or Material | Gold/Iridium/Osmium/Palladium/Platinum/Rhodium/Ruthenium/Tellurium |
| CAS | 7440-57-5, 7439-88-5, 4-2-7440, 5-3-7440, 6-4-7440, 7440-16-6 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Gold/Iridium/Osmium/Palladium/Platinum/Rhodium/Ruthenium/Tellurium |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HCl |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Gold 100 μg/mL/Iridium 100 μg/mL/Osmium 100 μg/mL/Palladium 100 μg/mL/Platinum 100 μg/mL/Rhodium 100 μg/mL/Ruthenium 100 μg/mL/Tellurium 100 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?