missing translation for 'onlineSavingsMsg'
Learn More

SLV-2436, MedChemExpress

Product Code. 16410877
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16410877

Brand: Medchem Express HY112113

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SLV-2436 is a highly potent and ATP-competitive inhibitor of MNK1 and MNK2 with IC50s of 10.8 nM and 5.4 nM, respectively.

SLV-2436 is a highly potent and ATP-competitive inhibitor of MNK1 and MNK2 with IC50s of 10.8 nM and 5.4 nM, respectively.

Specifications

Chemical Name or Material SLV-2436
Physical Form Solid
CAS 2095704-43-9
Color Brown
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C19H15ClN4O
For Use With (Application) Cancer-Kinase/protease
Synonym SEL201-88 SEL-201
Solubility Information DMSO : ≥ 100 mg/mL (285.06 mM)
SMILES O=C1C=CC(C2=CC3=C(C=C2)C(N)=NN3)=CN1CC4=CC=CC(Cl)=C4
Molecular Weight (g/mol) 350.8
Formula Weight 350.8
Percent Purity 98.47%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.