Learn More
3M™ Safety-Walk™ Slip-Resistant General Purpose Tapes and Treads, Series 613
Click to view available options
Dimensions (L x W):
18 x 0.051 m
18 x 0.152 m
18.3 x 0.025 m
18.3 x 0.102 m
18.3 x 0.305 m
18.3 x 0.610 m
Unit Size:
1 roll
2 rolls
4 rolls
Description
- Diagonal black/yellow striped tape provides early warning of slip prone areas
- Extremely visible, anti-slip tape alerts guests or workers to a potential hazard
- High quality abrasive particles to ensure long-lasting friction and visibility
- High performance pressure-sensitive adhesive for easy application
- NFSI High Traction Certification
Specifications
Specifications
| Dimensions (L x W) | 18.3 x 0.305 m |
| For Use With (Application) | Warning of Slip Prone Areas |
| Length (English) | 60 ft. |
| Length (Metric) | 18.3 m |
| Packaging Quantity | 1 Ea. |
| Width (English) | 12 in. |
| Width (Metric) | 0.305 m |
| Adhesive Type | Pressure Sensitive Adhesive |
| Color | Black and Yellow Striped |
| Type | Floor Tape |