Description
- Product Type: Slim T™ Mop Cover
- Length: 35 cm (14 in.)
- Cleaning Surface Fabric: PolySorb™ (Textured Polyester)
- Interior Layer: None (No sponge/Foam)
- Fasteners: Stainless Steel
- For Use With (Equipment): T-Mop™ Adapter (QDT, QPT, and QPWT Series)
Specifications
Specifications
| Dimensions | 6 x 35 cm (2.5 x 14 in.) |
| Product Type | Mop Cover |
| Head Type | Round |
| Color | White |
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Quantity | 12/Cs. (Individually Bagged) |
| Certifications/Compliance | ISO 9001 Certified Facility |
| Color-coded | No |
| Show More |