Learn More
Hecht Karl™ Slides
Click to view available options
Well Count:
1
2
3
Unit Size:
Each
Pack of 50
Description
- Microscope slides/plates with one or more cavities.
- Ground edges.
Specifications
Specifications
| Length (Metric) | 76 mm |
| Width (Metric) | 26 mm |
| No. per Pack | 20 |
| Edges | Ground |
| Thickness (Metric) | 3 to 4 mm |
| Type | Microscope Slide |
| Well Count | 1 |
| Coating | No Surface Treatment |
| Description | 22mm cavity diameter, 1.75mm cavity depth |
| Dimensions (L x W) | 76 x 26 mm |