Learn More
Epredia™ Slide Baskets and Hangers for Epredia™ ClearVue™ Coverslipper
Use these slide baskets and hangers with the Epredia™ ClearVue™ Coverslipper to make it compatible with any automated slide stainer.
161.00 EUR - 178.00 EUR
Specifications
Product Type | Coverslipper Basket/Hanger |
---|---|
For Use With (Equipment) | Epredia ClearVue Coverslipper |
Quantity | 5 Pk. |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
12628196
|
Epredia™
A79210064 |
Gemini-compatible baskets, black sliders |
167.00 EUR
Pack of 5 |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12638196
|
Epredia™
A79210065 |
Gemini-compatible basket, white sliders |
178.00 EUR
Pack of 5 |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12648196
|
Epredia™
A79210066 |
Sakura DRS 2000-compatible basket, black sliders |
161.00 EUR
Pack of 5 |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12688196
|
Epredia™
A79210072 |
Leica baskets, black sliders |
162.00 EUR
Pack of 5 |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Allow equipment compatibility with a variety of different stainers
- Can be loaded in mix-or-match fashion
- Sold in sets of five
Specifications
Coverslipper Basket/Hanger | |
Epredia ClearVue Coverslipper | |
5 Pk. |