missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC7A7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113527
This item is not returnable.
View return policy
Description
SLC7A7 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
The protein encoded by this gene is the light subunit of a cationic amino acid transporter. This sodium-independent transporter is formed when the light subunit encoded by this gene dimerizes with the heavy subunit transporter protein SLC3A2. This transporter is found in epithelial cell membranes where it transfers cationic and large neutral amino acids from the cell to the extracellular space. Defects in this gene are a cause of lysinuric protein intolerance (LPI). Several transcript variants encoding the same protein have been found for this gene.
Specifications
| SLC7A7 | |
| Polyclonal | |
| Unconjugated | |
| SLC7A7 | |
| AI790233; amino acid transporter SLC7A7 y+LAT1; LAT3; LPI; monocyte amino acid permease 2; MOP-2; my+lat1; Slc7a7; solute carrier family 7 (amino acid transporter light chain, y+L system), member 7; solute carrier family 7 (cationic amino acid transporter, y+ system), member 7; solute carrier family 7 member 7; y(+)L-type amino acid transporter 1; Y+L amino acid transporter 1; Y+L amino acid transporter 1; solute carrier family 7 member 7; y+LAT1; y+LAT-1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 20540, 9056 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9UM01, Q9Z1K8 | |
| SLC7A7 | |
| Recombinant Human Y+L amino acid transporter 1 protein (325-382AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction