missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SLC6A14 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-27859-50ul
This item is not returnable.
View return policy
Description
SLC6A14 Polyclonal antibody specifically detects SLC6A14 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| SLC6A14 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA 1:40000, Immunohistochemistry-Paraffin 1:100-300 | |
| Amino acid transporter ATB0+, amino acid transporter B0+, BMIQ11, OBX, sodium- and chloride-dependent neutral and basic amino acid transporter B(0+), solute carrier family 6 (amino acid transporter), member 14, solute carrier family 6 (neurotransmitter transporter), member 14, Solute carrier family 6 member 14 | |
| Synthesized peptide derived from the C-terminal region of human SLC6A14. (Uniprot #: Q9UN76) | |
| 50 μL | |
| Cancer, Endocrinology, Signal Transduction | |
| 11254 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, 50% glycerol, 0.5% BSA | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction