missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC4A10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-82594
This item is not returnable.
View return policy
Description
SLC4A10 Polyclonal specifically detects SLC4A10 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SLC4A10 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| sodium bicarbonate cotransporter-like, member 10, sodium bicarbonate transporter-like, member 10, sodium-driven chloride bicarbonate exchanger, Solute carrier family 4 member 10, solute carrier family 4, sodium bicarbonate transporter, member 10 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 57282 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| SLC4A10 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:FELSEAYPINMHNDLELLTQYSCNCVEPHNPSNGTLKEWRESNISASDIIWENLT | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction